
N-Methylanthranilic acid ≥99.0% (by HPLC, titration analysis)
Catálogo # TCIAM0154-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:151.16 g/mol
Boiling Pt:80 °C (0,01 -rr)
Melting Pt:172 °C
EINECS:204-343-9
:GHS07
Storage Temperature:Ambient
CAS Number:119-68-6
Formula:C₈H₉NO₂
MDL Number:MFCD00002424