3-Aminobenzoic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:99-05-8
:
Formula:C₇H₇NO₂
MDL Number:MFCD00007795
:137.14 g/mol
Merck Index:14,00421
Boiling Pt:308 °C (1013 hPa)
Melting Pt:174 °C
Density:1,5105 g/cm³ (20 °C)
EINECS:202-724-4
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together