Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

Vanillin ≥98.0% (by GC, titration analysis)

Irritant
Supplier:   TCI
Vanillin ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Vanillin ≥98.0% (by GC, titration analysis)

2 Options Available Below

Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:14,09932
  • Boiling Pt:284°C
  • Melting Pt:∼82 °C
  • Density:1,056 g/cm³ (20 °C)
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    80.5 to 83.5 deg-C
  • Solubility in EtOH:
    almost transparency
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 32,00 €
    32,00 €
    1 * 25 g
       32,00 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 102,00 €
    102,00 €
    1 * 500 g
       102,00 €