2-Bromo-5-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:22921-68-2
:
Formula:C₈H₇BrO₃
MDL Number:MFCD00020214
:231.05 g/mol
EINECS:245-329-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:159.0 to 163.0 deg-C
Specification Test Results
Frequently Bought Together