n-Dotriacontane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:544-85-4
:
Formula:H₃C(CH₂)₃₀CH₃
MDL Number:MFCD00009411
:450.88 g/mol
Boiling Pt:220°C
Density:0,7798 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-881-5
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:69.0 to 72.0 deg-C
- Solubility in hot Toluene:almost transparency
Specification Test Results
Frequently Bought Together