Order Entry
Portugal
ContactUsLinkComponent
Decyl acetate ≥98.0% (by GC)
Decyl acetate ≥98.0% (by GC)
  TCIAA1366-25ML
 :  TCI
 :  
undefined
Decyl acetate ≥98.0% (by GC)
  TCIAA1366-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :200.32 g/mol
 :248°C
 :203-942-2
 :112-17-4
 :C₁₂H₂₄O₂
 :MFCD00026538