3-Hexanone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:589-38-8
:
Formula:C₆H₁₂O
MDL Number:MFCD00009402
Boiling Pt:123
Molecular Weight:100.16 g/mol
Melting Pt:-55.4
Flash Pt:14°C
Storage Temperature:Ambient
EINECS:209-645-4
UN:1224
ADR:3 II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together