Order Entry
Portugal
ContactUsLinkComponent
 
 
4-Isopropyl-m-cresol ≥99.0% (by GC)
 :  TCI
 
undefined
 
4-Isopropyl-m-cresol ≥99.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    110.0 to 113.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

Frequently Bought Together