Order Entry
Portugal
ContactUsLinkComponent
1-Naphthalenesulphonyl chloride ≥98.0% (by titrimetric analysis)
Corrosive
1-Naphthalenesulphonyl chloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
1-Naphthalenesulphonyl chloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    66.0 to 70.0 deg-C

Specification Test Results

Frequently Bought Together