
4-Methylpyrocatechol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:452-86-8
:
Formula:C₇H₈O₂
MDL Number:MFCD00002205
:124.14 g/mol
Boiling Pt:117°C
EINECS:207-214-5
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:65.0 to 68.0 deg-C
Specification Test Results
Frequently Bought Together