Order Entry
Portugal
ContactUsLinkComponent
 
 
(Ethyl)triphenylphosphonium iodide ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
 
(Ethyl)triphenylphosphonium iodide ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    168.0 to 172.0 deg-C
  • Solubility in Methanol:
    within almost transparency

 

Frequently Bought Together