

Silver hexafluoroantimonate ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:26042-64-8
:
Formula:AgF₆Sb
MDL Number:MFCD00003401
:343.62
Density:4,65 to 4.85 g/cm³ (20 °C)
EINECS:247-429-1
UN:1759
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Precipitation Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together