

Benzyl(methyl)amine ≥98.0% (by GC, titration analysis)
: TCI
:103-67-3
:
:C₆H₅CH₂NHCH₃
:MFCD00008289
:121.18 g/mol
:186°C
:–55 °C
:0,94 g/cm³ (20 °C)
:77°C
:Ambient
:203-133-4
:2735
:8,II
:GHS05
:GHS08
- Appearance:Colorless to Light yellow to Red clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together