Zinc bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7699-45-8
:
Formula:Br₂Zn
MDL Number:MFCD00011294
Merck Index:14,01029
Boiling Pt:697°C
Melting Pt:394°C
Density:4,22 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-718-4
ADR:8,III
:225.2 g/mol
UN:3260
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together