trans-2-Methylcyclohexanol ≥98.0% (by GC)
Catalog # TCIAM0352-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:114.19 g/mol
Boiling Pt:167°C
Melting Pt:–41 °C
Density:0,9247 g/cm³ (20 °C)
Flash Pt:59°C
EINECS:231-186-3
UN:2617
ADR:3,III
:GHS02
:GHS07
CAS Number:7443-52-9
Formula:C₇H₁₄O
MDL Number:MFCD00064168