Calcium propionate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4075-81-4
:
Formula:(CH₃CH₂COO)₂Ca
MDL Number:MFCD00036137
:186.22 g/mol
Merck Index:14,01698
Melting Pt:> 300 °C
EINECS:223-795-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
- Solubility in Water:very faint turbidity
Specification Test Results
Related Information
Frequently Bought Together