



Dichloroacetic acid (DCA) ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:79-43-6
:
Formula:C₂H₂Cl₂O₂
MDL Number:MFCD00004223
Merck Index:14,03050
Boiling Pt:193°C
Melting Pt:∼–4 °C
Density:1,563 g/cm³ (20 °C)
Flash Pt:170°C
UN:1764
ADR:8,II
:128.94 g/mol
EINECS:201-207-0
:GHS05
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together