2-Methyl-2-pentanol ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:590-36-3
:
Formula:C₆H₁₄O
MDL Number:MFCD00004485
:102.18 g/mol
Boiling Pt:122°C
Melting Pt:–107 °C
Density:0,8262 g/cm³ (20 °C)
Flash Pt:30°C
Storage Temperature:Ambient
EINECS:209-681-0
UN:2560
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together