

2,4-Dimethyl-3-pentanol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:600-36-2
:
Formula:C₇H₁₆O
MDL Number:MFCD00008921
:116.2 g/mol
Boiling Pt:136°C
Melting Pt:–70 °C
Density:0,8445 g/cm³ (0 °C)
Flash Pt:37°C
EINECS:209-993-7
UN:1987
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together