3-Methoxyphenyl isocyanate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:18908-07-1
:
Formula:C₈H₇NO₂
MDL Number:MFCD00002019
:149.15 g/mol
Boiling Pt:94 °C (1013 hPa)
Density:1,13 g/cm³ (20 °C)
Flash Pt:96°C
Storage Temperature:Refrigerator
EINECS:242-658-3
UN:2206
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together