Order Entry
Portugal
ContactUsLinkComponent
 
2-Benzylphenol ≥97.0% (by GC)
 :  TCI
 
undefined
2-Benzylphenol ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Brown powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    51.0 to 54.0 deg-C

 

Frequently Bought Together