
4-Bromo-o-cresol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2362-12-1
:
Formula:C₇H₇BrO
MDL Number:MFCD00055435
Boiling Pt:88°C
:187.04 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:64.0 to 68.0 deg-C
Specification Test Results
Frequently Bought Together