Thymol blue
Supplier: TCI
About this item
Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-61-9
- Formula:C₂₇H₃₀O₅S
- MDL Number:MFCD00005869
- Molecular Weight:466.6 g/mol
- Merck Index:14,09400
- Melting Pt:221 °C
- Storage Temperature:Ambient
- EINECS:200-973-3
2 Options Available Below
Product Details & Documents
Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-61-9
- Formula:C₂₇H₃₀O₅S
- MDL Number:MFCD00005869
- Molecular Weight:466.6 g/mol
- Merck Index:14,09400
- Melting Pt:221 °C
- Storage Temperature:Ambient
- EINECS:200-973-3
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to Brown to Dark green powder to crystaline
- Absorbance:min. 0.270 (10mg/L, pH 7.6, 431.0 to 437.0 nm)
- Solubility in Sodium hydroxide solution:almost transparency
- Transition interval (pH):(red)1.2 to 2.8 (yellow), (yellow)8.0 to 9.6 (blue)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



