Screw top vials and closures, 13 mm
Fornecedor: Agilent
Certificados
Sobre este item
These screw top vials with precision-formed screw threads of 13-425 are ideal for consistent secure seal and can be used as wash, waste, or storage vials.
Screw caps with 13 mm septa
Informações do Produto
Frequently Bought Together