Order Entry
Portugal
ContactUsLinkComponent
Organic reference standard, Hymecromone (4-Methylumbelliferone, 7-Hydroxy-4-methylcoumarin) 100 µg/ml in acetonitrile, CIL
Organic reference standard, Hymecromone (4-Methylumbelliferone, 7-Hydroxy-4-methylcoumarin) 100 µg/ml in acetonitrile, CIL
  PROOCILULM73091.2
 :  
Organic reference standard, Hymecromone (4-Methylumbelliferone, 7-Hydroxy-4-methylcoumarin) 100 µg/ml in acetonitrile, CIL
  PROOCILULM73091.2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    90-33-5
  • Description:
    Hymecromone (4-Methylumbelliferone, 7-Hydroxy-4-methylcoumarin) 100 µg/ml in acetonitrile
  • Formula:
    C₁₀H₈O₃
  • Description:
    Hymecromone (4-Methylumbelliferone, 7-Hydroxy-4-methylcoumarin)