Order Entry
Portugal
ContactUsLinkComponent
Chloramphenicol 97.0-103.0%, Ultra Pure Grade
Chloramphenicol 97.0-103.0%, Ultra Pure Grade
Catalog # 0230-EU-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Chloramphenicol 97.0-103.0%, Ultra Pure Grade
Catalog # 0230-EU-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Chromatographic Purity:
    PASS
  • Crystallinity:
    PASS
  • Identification:
    Pass
  • Melting Range:
    149 – 153 °C
  • pH (2.5%, Water) @ 25 °C:
    4.5 – 7.5
  • Purity:
    97.0 – 103.0 %
  • Specific Rotation:
    +17.0 – +20.0°

Specification Test Results

About this item

 :
 :323.13 g/mol
Merck Index:13,02087
Boiling Pt:645 °C (1013 hPa)
Melting Pt:150 °C
Density:1.49 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-287-4
 :GHS08
CAS Number:56-75-7
Formula:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MDL Number:MFCD00078159