
4,4'-Dinonyl-2,2'-bipyridyl ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:142646-58-0
:
Formula:C₂₈H₄₄N₂
MDL Number:MFCD00800915
:408.67 g/mol
Melting Pt:61…63 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together