
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-8, SB-8), Sigma-Aldrich®
Catalog # SIALO6626-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
:GHS07
Storage Temperature:Ambient
CAS Number:15178-76-4
Formula:C₁₃H₂₉NO₃S
MDL Number:MFCD00036912