TBPP (tris(2,3-dibromopropyl)phosphate)
Catalog # EHERC17894340
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
EINECS:204-799-9
UN:3082
ADR:9,III
CAS Number:126-72-7
Formula:C₉H₁₅Br₆O₄P