Order Entry
Portugal
ContactUsLinkComponent
Supelco® Organic Reference Standard, Malathion Solution
Supelco® Organic Reference Standard, Malathion Solution
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Malathion Solution
Supplier:  Merck

Specifications

  • CAS No.:
    121-75-5
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H225 - H304 - H315 - H336 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00036233
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₀H₁₉O₆PS₂
  • Molecular weight:
    330,36
  • Flash point:
    –18 °C - closed cup

Specifications

Frequently Bought Together