Hydrofluoric acid ≥70% in pyridine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7664-39-3
:
Formula:HF
MDL Number:MFCD00011346
:20.01 g/mol
:M-Clarity™ quality level = MQ200
EINECS:231-634-8
Frequently Bought Together