(±)-1,2,3,4-Tetrahydro-1-naphthylamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2217-40-5
:
Formula:C₁₀H₁₃N
MDL Number:MFCD00001740
:M-Clarity™ quality level = MQ100
:147.22 g/mol
EINECS:218-712-7
Frequently Bought Together