Bismuth subsalicylate ≥96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Bismuth(III) subsalicylate, 2-Hydroxy-4H-benzo[d][1,3,2]dioxabismin-4-one
CAS Number:14882-18-9
:
Formula:C₇H₅BiO₄
MDL Number:MFCD00085368
Molecular Weight:362.09 g/mol
EINECS:238-953-1
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together