Divinylbenzene (mixture of isomers) stabilised, Technical Grade, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1321-74-0
:
Formula:C₁₀H₁₀
MDL Number:MFCD01778656
:130.19 g/mol
:M-Clarity™ quality level = MQ100
:Stabilised with 4-tert-butylpyrocatechol (TBC)
EINECS:215-325-5
Frequently Bought Together