Order Entry
Portugal
ContactUsLinkComponent
Cyclopentyl methyl ether (CPME) ≥99.5% (by GC) stabilised
Cyclopentyl methyl ether (CPME) ≥99.5% (by GC) stabilised
Supplier:  TCI
Certificates
undefined
Cyclopentyl methyl ether (CPME) ≥99.5% (by GC) stabilised
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.5 %

Specification Test Results

Frequently Bought Together