2-Naphthol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:135-19-3
:
Formula:C₁₀H₈O
MDL Number:MFCD00004067
Merck Index:12,06471
Boiling Pt:286 °C (1013 hPa)
Melting Pt:121.6 °C
Density:1.217 g/cm³ (20 °C)
Flash Pt:153 °C
EINECS:205-182-7
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
:144.17 g/mol
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:1 g/l (20 °C)
- Melting Point:121.6 °C
- Molar Mass:144.17 g/mol
- Bulk Density:300 kg/m3
- Boiling Point:294.8 °C (1013 hPa)
- Vapor Pressure:- 0.1 hPa (30 °C)
- Flash Point:153 °C
- Density:1.27 g/cm3 (20 °C)
- Ignition Point:430 °C
Specification Test Results
Frequently Bought Together