Diphosphorus pentoxide, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1314-56-3
:
Formula:P₂O₅
MDL Number:MFCD00011440
:141.94 g/mol
Merck Index:13,07441
Melting Pt:562 °C
Density:2.253 g/cm³ (20 °C)
EINECS:215-236-1
ADR:8,II
:M-Clarity™ quality level = MQ200
UN:1807
:GHS05
Storage Temperature:Ambient
Frequently Bought Together