Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
di-Phosphorus pentoxide
Diphosphorus pentoxide ≥98%
Diphosphorus pentoxide ≥98%
Diphosphorus pentoxide ≥98%
A13348.0E
:
Thermo Fisher Scientific
:
Diphosphorus pentoxide ≥98%
A13348.0E
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
2.500 g
Seq:
3795
:
1314-56-3
:
:
P₂O₅
:
MFCD00011440
:
141.94 g/mol
:
14,7355
:
360 °C (subl.)
:
340 °C
:
2.39
:
Ambient
:
215-236-1
:
1807
:
8,II
:
GHS05