Diphosphorus pentoxide 98+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1314-56-3
:
Formula:P₂O₅
MDL Number:MFCD00011440
:141.94 g/mol
Merck Index:13,07441
Melting Pt:340…360 °C (subl.)
Density:2,253 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-236-1
UN:1807
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White Powder
- Titration with NaOH:≥98.0 %
- Ammonium (NH4):≤100 ppm
- Heavy metals (as Pb):≤100 ppm
- Phosphorus trioxide (P2O3):Passes test (limit about 0.02 %)
- Insoluble matter:≤0.02 %
Specification Test Results
Frequently Bought Together