GL Quick tube connectors, BOLA
Supplier: Bohlender
Certificates
About this item
Two-part quick connector is made of PFA, with two GL threads. When disconnected, the flow is interrupted by means of built-in non-return valves.
- Suitable for pressure up to max. 6 bar
- Universal chemical resistance
- Maximum temperature up to 250 °C
Frequently Bought Together