Sodium fluoride, Optical Grade
Catalog # 11023.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:41.99 g/mol
Merck Index:14,8618
Boiling Pt:1704 °C
Melting Pt:993 °C
Density:2.78
Storage Temperature:Ambient
EINECS:231-667-8
UN:1690
ADR:6.1,III
:GHS06
:GHS07
CAS Number:7681-49-4
Formula:NaF
MDL Number:MFCD00003524