Potassium bromide, ultra dry ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-02-3
:
Formula:KBr
MDL Number:MFCD00011358
Merck Index:14,7618
Boiling Pt:1435 °C
Melting Pt:730 °C
Density:2.75
:ampouled under argon
:119 g/mol
EINECS:231-830-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together