
Paraldehyde
Supplier: Merck
Certificates
About this item
CAS Number:123-63-7
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00036208
:132.16 g/mol
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:204-639-8
UN:1264
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Solubility:120 g/l (13 °C)
- Melting Point:12 °C
- Molar Mass:132.15 g/mol
- Boiling Point:124 °C (1013 hPa)
- Vapor Pressure:13.3 hPa (20 °C)
- Flash Point:27 °C
- Refractive Index:1.4049 (20 °C)
- Explosion Limit:1.3 - 17.0 %(V)
- Density:0.99 g/cm3 (20 °C)
- Ignition Point:235 °C
Specification Test Results
Frequently Bought Together