Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Paraldehyde
Paraldehyde
Paraldehyde
EHERC15848200
:
EHRENSTORFER
:
Paraldehyde
EHERC15848200
:
EHRENSTORFER
:
Pack type:
Glass bottle
Pk:
5 mL
Seq:
2210
:
:
132.16 g/mol
:
204-639-8
:
1264
:
3,III
:
123-63-7
:
C₆H₁₂O₃
:
MFCD00036208