Paraldehyde 99.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-63-7
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00036208
:132.16 g/mol
Melting Pt:12 °C
Density:0.99 g/cm³
Flash Pt:24 °C
Storage Temperature:Ambient
EINECS:204-639-8
UN:1264
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4035 to 1.4055 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥99.4 %
- Free acid (CH3COOH):≤0.3 % (titr.)
Specification Test Results
Frequently Bought Together