

Benzotrifluoride for synthesis, Sigma-Aldrich®
Catalog # 8.41554.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Hill Formula:C₇H₅F₃
- Molar Mass:146.11 g/mol
- Boiling point:100 - 103 °C (1013 hPa)
- Density:1.197 g/cm³ (20 °C)
- Explosion limit:1.4 - 9.3 %(V)
- Flash point:12 °C
- Ignition temperature:650 °C
- Melting Point:-29 °C
- Vapor pressure:53 hPa (25 °C)
- Solubility:0.45 g/l (20 °C) Hydrolysis
- LD 50 oral:LD50 Rat 15000 mg/kg
- LD 50 dermal:LD50 Rat 2000 mg/kg
- Storage class:3 Flammable liquids
- WGK:WGK 1 slightly water endangering
- R Phrase:R 11-51/53
- S Phrase:S 16-23-61
Specification Test Results
About this item
:
:146.11 g/mol
Merck Index:13,01111
Boiling Pt:101 °C (1013 hPa)
Melting Pt:–29 °C
Density:1.37 g/cm³ (20 °C)
Flash Pt:12 °C
EINECS:202-635-0
UN:2338
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS09
Storage Temperature:Ambient
CAS Number:98-08-8
Formula:C₇H₅F₃
MDL Number:MFCD00000372