Order Entry
Portugal
Orders LinkContactUsLinkComponent
Clamps, micro
Clamps, micro
Supplier:   BOCHEM
Clamps, micro
Supplier:  BOCHEM
Certificates
Clamps, micro
3 Options Available Below
Product Details & Documents
Micro clamps made of stainless steel.
  • Fingers are covered with either a layer of cork or with silicone coating to protect clamped items
  • With round or angled jaws
Product Family Options
Product Information
  • Material
    Spanning width
    Length
    Availability
    Price
  • BOCH5405
    Clamps, micro, finger with cork, angled jaws
    Clamps, micro, finger with cork, angled jaws
    Stainless steel 18/10
    15 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 unid.
    1 * 1 unid. - 28,50 €
    28,50 €
    1 * 1 unid.
       28,50 €
  • BOCH5415
    Clamps, micro, finger with silicone coating
    Clamps, micro, finger with silicone coating
    Stainless steel 18/10
    0 - 20
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 unid.
    1 * 1 unid. - 37,90 €
    37,90 €
    1 * 1 unid.
       37,90 €
  • BOCH5400
    Clamps, micro, finger with cork, round jaws
    Clamps, micro, finger with cork, round jaws
    Stainless steel 18/10
    15 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 unid.
    1 * 1 unid. - 28,50 €
    28,50 €
    1 * 1 unid.
       28,50 €