About this item
Braid reinforced for increased pressure resistance, TYGON® 3370 I.B. silicone hose is frequently specified in the most demanding applications requiring sanitary transfer of fluids.
- Consistently smooth inner surface limits particle entrapment
- Platinum cured to minimize extractables
- Tough braid reinforcement permits use under elevated working pressures
- Withstands repeated CIP and SIP cleaning and sterilization
Its smooth inner surface reduces the risk of particle entrapment and inhibits excessive residue and microscopic bacterial build-up; cleaning and sterilization cycles may become more effective as a result. Additionally, an improvement in fluid flow characteristics may occur from the reduced surface area and lowered absorption of fluids to the wall.
TYGON® 3370 I.B. can easily withstand repeated SIP and CIP cleaning and sterilization cycles, making it ideal for repeat-use applications. Its flexibility, durability, and chemical and temperature resistance provide a unique combination of characteristics required in many pharmaceutical, biotechnology, laboratory, food, beverage, cosmetic, environmental, and industrial applications.
Temperature resistance: −80 to +160 °C
Hardness (Shore durometer A): 70
Density: 1,18