
trans-4-Methylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13064-83-0
:
Formula:C₈H₁₄O₂
MDL Number:MFCD00074943
:142.2 g/mol
Boiling Pt:245°C
EINECS:235-959-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
- Melting point:110.0 to 114.0 deg-C
Specification Test Results
Frequently Bought Together