Order Entry
Portugal
Orders LinkContactUsLinkComponent
Scratcher for sampling
Scratcher for sampling
Supplier:   BURKLE
Scratcher for sampling
Supplier:  BURKLE
Certificates
Scratcher for sampling
Scratcher for sampling
3 Options Available Below
Product Details & Documents
The scratcher has particularly been designed for sampling cohesive or caked substances.
  • Electro-polished stainless steel
For non pourable substances.
Product Family Options
Product Information
  • Material
    Length
    Ø
    Volume
    Availability
    Price
  • Stainless steel V4A
    1000 mm
    25 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 1 unid.
    1 * 1 unid. - 1 350,00 €
    1 350,00 €
    1 * 1 unid.
       1 350,00 €
  • Stainless steel V4A
    1000 mm
    25 mm
    5 ml
    Log in for availability
    Log in for availability
    1 * 1 unid.
    1 * 1 unid. - 1 290,00 €
    1 290,00 €
    1 * 1 unid.
       1 290,00 €
  • Stainless steel V4A
    1000 mm
    25 mm
    25 ml
    Log in for availability
    Log in for availability
    1 * 1 unid.
    1 * 1 unid. - 1 420,00 €
    1 420,00 €
    1 * 1 unid.
       1 420,00 €